missing translation for 'onlineSavingsMsg'
Learn More

L-Isoserine, 98%

Catalog No. AAH60945MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH60945MD 250 mg
AAH6094503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH60945MD Supplier Thermo Scientific Chemicals Supplier No. H60945MD
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 632-13-3 | C3H7NO3 | 105.09 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Slightly soluble in water.

Notes
Incompatible with strong oxidizing agents. Store under room temperature. Moisture sensitive.

Chemical Identifiers

CAS 632-13-3
Molecular Formula C3H7NO3
Molecular Weight (g/mol) 105.09
MDL Number MFCD03428326
InChI Key BMYNFMYTOJXKLE-REOHCLBHSA-N
Synonym l-isoserine, s-isoserine, s-3-amino-2-hydroxypropanoic acid, s-3-amino-2-hydroxypropionic acid, 2s-3-amino-2-hydroxypropanoic acid, propanoic acid, 3-amino-2-hydroxy-, 2s, s-2-hydroxy-, a-alanine, s-2-hydroxy-beta-alanine, s-2-hydroxy-b-alanine
PubChem CID 6999797
SMILES [NH3+]C[C@H](O)C([O-])=O

Specifications

Melting Point 197°C to 198°C
Solubility Information Slightly soluble in water.
IUPAC Name (2S)-3-amino-2-hydroxypropanoic acid
Quantity 250 mg
Formula Weight 105.09
Percent Purity 98%
Sensitivity Moisture sensitive
Chemical Name or Material L-Isoserine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only