missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucinol 97.0+%, TCI America™

Catalog No. I04621G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I04621G 1 g
I04625G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I04621G Supplier TCI America Supplier No. I04621G

Chemical Identifiers

CAS 24629-25-2
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00004731
InChI Key VTQHAQXFSHDMHT-NTSWFWBYSA-O
Synonym l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
PubChem CID 2724027
IUPAC Name (2S,3S)-1-hydroxy-3-methylpentan-2-aminium
SMILES CC[C@H](C)[C@H]([NH3+])CO

Specifications

Melting Point 42°C
Color White-Yellow
Boiling Point 97°C
Quantity 1 g
Formula Weight 117.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material L-Isoleucinol