accessibility menu, dialog, popup

UserName

CAS RN 24629-25-2

IUPAC Name: (2S,3S)-1-hydroxy-3-methylpentan-2-aminium
Synonyms: (2S,3S)-1-hydroxy-3-methylpentan-2-aminium
Molecular Weight (g/mol): 118.2
Molecular Formula: C6H16NO
InChi Key: VTQHAQXFSHDMHT-NTSWFWBYSA-O
SMILES: CC[C@H](C)[C@H]([NH3+])CO
Boiling Point 
  • (2)
  • (1)
  • (1)
Molecular Weight (g/mol) 
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)