missing translation for 'onlineSavingsMsg'
Learn More

L(+)-Isoleucinol, 97%

CAS: 24629-25-2 | C6H16NO | 118.20 g/mol

$117.84 - $117.84

Chemical Identifiers

CAS 24629-25-2
Molecular Formula C6H16NO
Molecular Weight (g/mol) 118.20
MDL Number MFCD00004731
InChI Key VTQHAQXFSHDMHT-NTSWFWBYSA-O
Synonym l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
PubChem CID 2724027
IUPAC Name (2S,3S)-2-amino-3-methylpentan-1-ol
SMILES CC[C@H](C)[C@H]([NH3+])CO
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC183940010
SDS View Documents
Thermo Scientific Chemicals
183940010
1 g Glass bottle
Each for $117.84
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

24629-25-2
118.20
VTQHAQXFSHDMHT-NTSWFWBYSA-O
2724027
CC[C@H](C)[C@H]([NH3+])CO
C6H16NO
MFCD00004731
l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
(2S,3S)-2-amino-3-methylpentan-1-ol

Specifications

24629-25-2
White to Yellow
100°C
97%
C6H16NO
C2H5CH(CH3)CH(NH2)CH2OH
1 g
+ 4.5
VTQHAQXFSHDMHT-NTSWFWBYSA-O
(2S,3S)-2-amino-3-methylpentan-1-ol
2724027
97%
L(+)-Isoleucinol, 97%
36°C to 39°C
97°C (14.0 mmHg)
Authentic
Glass bottle
1.4570 to 1.46
MFCD00004731
+ 4.50 (20.00°C c=1.6 C2H5OH)
l-isoleucinol, 2s,3s-2-amino-3-methylpentan-1-ol, s-+-isoleucinol, h-isoleucinol, l-+-isoleucinol, isoleucinol, 2s,3s-2-amino-3-methyl-pentan-1-ol, h-ile-ol, 1-pentanol, 2-amino-3-methyl-, 2s,3s
CC[C@H](C)[C@H]([NH3+])CO
118.20
117.19
Viscous Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present a

GHS Signal Word: Warning

EINECSNumber : 246-371-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only