missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111015376
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111015376 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111015376 Supplier Merck Emd Millipore Supplier No. PHR10991G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

L-Isoleucine is an isomer of L-leucine and an essential amino acid. It is synthesized from threonine and is a branched-chain hydrophobic amino acid.

Specifications

CAS 73-32-5
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula C2H5 CH(CH3)CH(NH2)CO2 H
MDL Number MFCD00064222
Melting Point 288°C
Synonym (2 S,3 S)-2-Amino-3-methylpentanoic acid
Type Certified Reference Material
Quantity 1 g
Formula Weight 131.17
Grade Certified Reference Material
Show More Show Less