missing translation for 'onlineSavingsMsg'
Learn More

L-Isoleucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1784714

 View more versions of this product

Catalog No. AAA1784714


Only null left

Description

Description

L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Isoleucine methyl ester used for chemical synthesis. E.g. It can also be used to produce 3-methyl-2-(4-nitro-imidazol-1-yl)-pentanoic acid methyl ester. It also finds its use as a pharmaceutical intermediate.

Solubility
Soluble in methanol (50 mg/ml clear, colorless solution).

Notes
Hygroscopic. Keep away from oxidizing agents, water/moisture. Store under inert atmosphere. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. In order to obtain a higher solubility, warm the tube at 37°C and shake it in the ultrasonic bath for a while.
Specifications

Specifications

98°C to 100°C
18598-74-8
MFCD00038911
l-isoleucine methyl ester hydrochloride, h-ile-ome.hcl, methyl l-isoleucinate hydrochloride, methyl l-isoleucinate hcl, methyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, 2s,3s-methyl 2-amino-3-methylpentanoate hydrochloride, l-isoleucine, methyl ester, hydrochloride, l-isoleucine methyl ester hydrochloride;h-ile-ome.hcl, h-lle-ome.hcl, h-lle-ome?cl
GGTBEWGOPAFTTH-GEMLJDPKSA-N
methyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
181.66
181.66
≥98%
L-Isoleucine methyl ester hydrochloride
C7H16ClNO2
3595132
Soluble in methanol (50mg/ml clear,colorless solution).
Cl.CC[C@H](C)[C@H](N)C(=O)OC
25 g
11171351
Hygroscopic

Chemical Identifiers

18598-74-8
181.66
GGTBEWGOPAFTTH-GEMLJDPKSA-N
11171351
Cl.CC[C@H](C)[C@H](N)C(=O)OC
C7H16ClNO2
MFCD00038911
l-isoleucine methyl ester hydrochloride, h-ile-ome.hcl, methyl l-isoleucinate hydrochloride, methyl l-isoleucinate hcl, methyl 2s,3s-2-amino-3-methylpentanoate hydrochloride, 2s,3s-methyl 2-amino-3-methylpentanoate hydrochloride, l-isoleucine, methyl ester, hydrochloride, l-isoleucine methyl ester hydrochloride;h-ile-ome.hcl, h-lle-ome.hcl, h-lle-ome?cl
methyl (2S,3S)-2-amino-3-methylpentanoate;hydrochloride
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 242-437-1

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only