missing translation for 'onlineSavingsMsg'
Learn More

L-α-Hydroxyglutaric acid-13C5 disodium salt, ≥98% (GC), MilliporeSigma™ Supelco™

$772.58 - $772.58

Specifications

Linear Formula 13C5H6 Na2O5
Synonym -2-Hydroxypentanedioic acid-1,2,3,4,5-13C5 disodium salt; Disodium -2-hydroxyglutarate-13C5
Type Analytical Standard
Formula Weight 197.04
Percent Purity ≥98% (GC)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
111003831
View Documents
Merck Emd Millipore
8055910MG
10 mg
Each for $772.58
Only null left
 
Description

Description

Specifications

Specifications

13C5H6 Na2O5
Analytical Standard
≥98% (GC)
-2-Hydroxypentanedioic acid-1,2,3,4,5-13C5 disodium salt; Disodium -2-hydroxyglutarate-13C5
197.04
Analytical Standard
Videos
SDS
Documents

Documents