missing translation for 'onlineSavingsMsg'
Learn More

L-Homophenylalanine, 98%

Catalog No. AAH2729503
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2729503 1 g
AAH2729506 5 g
AAH2729514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2729503 Supplier Thermo Scientific Chemicals Supplier No. H2729503

CAS: 943-73-7 | C10H13NO2 | 179.22 g/mol

L-Homophenylalanine is used as a precursor in the pharmaceutical industry for the production of angiotensin-converting enzyme (ACE) and R-(-)-Homophenylalanin-ethylester. It is also involved in the synthesis of NEPA [(S,S)-N-(1-Ethoxycarbonyl-3-phenylpropyl)alanine], which is a key intermediate to prepare the ACE-inhibitors: enalapril, delapril, quinapril and ramipril. It acts as an anti-tumor reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Homophenylalanine is used as a precursor in the pharmaceutical industry for the production of angiotensin-converting enzyme (ACE) and R-(-)-Homophenylalanin-ethylester. It is also involved in the synthesis of NEPA [(S,S)-N-(1-Ethoxycarbonyl-3-phenylpropyl)alanine], which is a key intermediate to prepare the ACE-inhibitors: enalapril, delapril, quinapril and ramipril. It acts as an anti-tumor reagent.

Solubility
Soluble in dilute aqueous acid.

Notes
Store in cool place. Incompatible with strong oxidizing agents, strong acids and bases.

Chemical Identifiers

CAS 943-73-7
Molecular Formula C10H13NO2
Molecular Weight (g/mol) 179.22
MDL Number MFCD00002619
InChI Key JTTHKOPSMAVJFE-VIFPVBQESA-N
Synonym l-homophenylalanine, homophenylalanine, s-2-amino-4-phenylbutanoic acid, s-2-amino-4-phenylbutyric acid, 2s-2-amino-4-phenylbutanoic acid, homophenylalanine, l, h-hophe-oh, +-2-amino-4-phenylbutyric acid, l-homophe
PubChem CID 2724505
ChEBI CHEBI:43103
SMILES N[C@@H](CCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point >300°C
Solubility Information Soluble in dilute aqueous acid.
IUPAC Name (2S)-2-amino-4-phenylbutanoic acid
Quantity 1 g
Formula Weight 179.22
Optical Rotation +45° (c=1 in 3N HCl)
Percent Purity 98%
Chemical Name or Material L-Homophenylalanine
EINECSNumber 213-403-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only