accessibility menu, dialog, popup

UserName

CAS RN 943-73-7

NH2(S)OHO

CAS RN 943-73-7

IUPAC Name: (2S)-2-amino-4-phenylbutanoic acid
Synonyms: D-homophenylalanine
Molecular Weight (g/mol): 179.22
Molecular Formula: C10H13NO2
InChi Key: JTTHKOPSMAVJFE-VIFPVBQESA-N
SMILES: N[C@@H](CCC1=CC=CC=C1)C(O)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)