missing translation for 'onlineSavingsMsg'
Learn More

L-Homoarginine Hydrochloride 97.0+%, TCI America™

Catalog No. H11721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H11721G 1 g
H11725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H11721G Supplier TCI America Supplier No. H11721G

Chemical Identifiers

CAS 1483-01-8
Molecular Formula C7H17ClN4O2
Molecular Weight (g/mol) 224.689
MDL Number MFCD00012622
InChI Key YMKBVNVCKUYUDM-JEDNCBNOSA-N
Synonym l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
PubChem CID 2723930
IUPAC Name (2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride
SMILES C(CCN=C(N)N)CC(C(=O)O)N.Cl

Specifications

Melting Point 206°C
Color White
Quantity 1 g
Formula Weight 224.69
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Homoarginine Hydrochloride