missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Homoarginine hydrochloride, 98%

CAS: 1483-01-8 | C7H16N4O2·HCl | 224.69 g/mol

$149.62 - $149.62

Chemical Identifiers

CAS 1483-01-8
Molecular Formula C7H16N4O2·HCl
Molecular Weight (g/mol) 224.69
MDL Number MFCD00012622
InChI Key YMKBVNVCKUYUDM-JEDNCBNOSA-N
Synonym l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
PubChem CID 2723930
IUPAC Name (2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride
SMILES C(CCN=C(N)N)CC(C(=O)O)N.Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC169090010
SDS View Documents
Thermo Scientific Chemicals
169090010
1 g Glass bottle
Each for $149.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1483-01-8
224.69
YMKBVNVCKUYUDM-JEDNCBNOSA-N
2723930
C(CCN=C(N)N)CC(C(=O)O)N.Cl
C7H16N4O2·HCl
MFCD00012622
l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
(2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride

Specifications

1483-01-8
White
98%
C7H16N4O2·HCl
MFCD00012622
+18.00 (20.00°C c=1,1N HCl)
l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
YMKBVNVCKUYUDM-JEDNCBNOSA-N
(2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride
2723930
98%
L(+)-Homoarginine hydrochloride, 99+%
213.0°C to 215.0°C
Authentic
Glass bottle
H2NC(=NH)NH(CH2)4CH(NH2)CO2H·HCl
1 g
+ 18.00
Solubility in water: 50mg/mL
C(CCN=C(N)N)CC(C(=O)O)N.Cl
224.69
224.69
Granular Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-045-6

SDS
Documents

Documents

RUO – Research Use Only