missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Homoarginine hydrochloride, 98%

Catalog No. AC169090010
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC169090010 1 g Glass bottle
AC169090050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC169090010 Supplier Thermo Scientific Chemicals Supplier No. 169090010
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 1483-01-8 | C7H16N4O2·HCl | 224.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1483-01-8
Molecular Formula C7H16N4O2·HCl
Molecular Weight (g/mol) 224.69
MDL Number MFCD00012622
InChI Key YMKBVNVCKUYUDM-JEDNCBNOSA-N
Synonym l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
PubChem CID 2723930
IUPAC Name (2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride
SMILES C(CCN=C(N)N)CC(C(=O)O)N.Cl

Specifications

Melting Point 213.0°C to 215.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula H2NC(=NH)NH(CH2)4CH(NH2)CO2H·HCl
Quantity 1 g
Specific Rotation Condition +18.00 (20.00°C c=1,1N HCl)
Specific Rotation + 18.00
Solubility Information Solubility in water: 50mg/mL
Formula Weight 224.69
Percent Purity 98%
Physical Form Granular Powder
Chemical Name or Material L(+)-Homoarginine hydrochloride, 99+%
Show More Show Less
EINECSNumber 216-045-6

RUO – Research Use Only