missing translation for 'onlineSavingsMsg'
Learn More

L-Homoarginine hydrochloride, 98%

Catalog No. p-7029873
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2738703 1 g
AAH2738706 5 g
AAH2738714 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH2738703 Supplier Thermo Scientific Chemicals Supplier No. H2738703

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1483-01-8 | C7H17ClN4O2 | 224.689 g/mol

An inhibitor of alkaline phosphatase isoenzymes. It is also an inhibitor of cellular transport of arginine by a sodium-independent high affinity y+ transporter. The substitution of homoarginine for arginine or lysine renders proteins resistant to proteolysis by trypsin. Arginine residues are often located at the active centers of proteins and enzymes. Replacing these arginines with homoarginine can elucidate protein function and structural requirements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An inhibitor of alkaline phosphatase isoenzymes. It is also an inhibitor of cellular transport of arginine by a sodium-independent high affinity y+ transporter. The substitution of homoarginine for arginine or lysine renders proteins resistant to proteolysis by trypsin. Arginine residues are often located at the active centers of proteins and enzymes. Replacing these arginines with homoarginine can elucidate protein function and structural requirements.

Solubility
Soluble in water(50 mg/mL).

Notes
Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated(2-7°C).

Chemical Identifiers

CAS 1483-01-8
Molecular Formula C7H17ClN4O2
Molecular Weight (g/mol) 224.689
MDL Number MFCD00012622
InChI Key YMKBVNVCKUYUDM-JEDNCBNOSA-N
Synonym l-homoarginine hydrochloride, s-2-amino-6-guanidinohexanoic acid hydrochloride, l +-homoarginine hydrochloride, h-homoarg-oh.hcl, l-homoarginine hcl, l-+-homoarginine hydrochloride, h-homoarg-oh, h-dl-hcit-oh, h-homoarg-oh hcl, h-har-oh hcl
PubChem CID 2723930
IUPAC Name (2S)-2-amino-6-(diaminomethylideneamino)hexanoic acid;hydrochloride
SMILES C(CCN=C(N)N)CC(C(=O)O)N.Cl

Specifications

Melting Point 213°C to 215°C
Beilstein 4016621
Solubility Information Soluble in water(50mg/mL).
Quantity 1 g
Formula Weight 224.69
Optical Rotation +18° (c=1 in 1N HCl)
Percent Purity ≥98%
Chemical Name or Material L-Homoarginine hydrochloride
EINECSNumber 216-045-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only