missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine, USP, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7157535
Change view
Click to view available options
Quantity:
25 g
100 g
1 kg
Packaging:
Poly Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603861 25 g Poly Bottle
18603862 100 g Poly Bottle
18603863 1 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18603861 Supplier Spectrum Chemical Mfg Cor Supplier No. H102125GM
Only null left

H1021, 71-00-1, C6H9N3O2

Spectrum™ Chemical L-Histidine, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
IUPAC Name 2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

CAS Max % 100%
Chloride 0.05%
Ignition Residue 0.4%
Loss on Drying 0.2%
Packaging Poly Bottle
Specific Rotation +12.6° to +14.0°
Sulfate 0.03%
Quantity 25 g
Percent Purity 98.5 to 101.5%
Grade USP