missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Histidine, USP, 98.5-101.5%, Spectrum™ Chemical
Description
Spectrum™ Chemical L-Histidine, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| CAS Max % | 100% |
| Chloride | 0.05% |
| Ignition Residue | 0.4% |
| Loss on Drying | 0.2% |
| Packaging | Poly Bottle |
| Specific Rotation | +12.6° to +14.0° |
| Sulfate | 0.03% |
| Quantity | 100 g |
| Percent Purity | 98.5 to 101.5% |
| Grade | USP |