missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Histidine Monohydrochloride Monohydrate, MP Biomedical
Supplier: MP Biomedicals Inc 0210195780
| Quantity | 100 g |
|---|
Chemical Identifiers
| C6H12ClN3O3 | |
| MFCD00151027 | |
| l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
| (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride |
Specifications
| L-Histidine Monohydrochloride Monohydrate | |
| White | |
| MFCD00151027 | |
| CMXXUDSWGMGYLZ-XRIGFGBMSA-N | |
| (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride | |
| 209.63 | |
| 209.6 |
| 5934-29-2 | |
| C6H12ClN3O3 | |
| l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate | |
| O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O | |
| 100 g | |
| 165377 | |
| Powder |
Safety and Handling
Recommended Storage : Store at Room Temperature (15 to 30°C).