missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine monohydrochloride monohydrate, Cell Culture Reagent

Catalog No. p-7042140
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6146509 10 g
AAJ6146522 100 g
AAJ6146536 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6146509 Supplier Thermo Scientific Chemicals Supplier No. J6146509
Only null left

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol

L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine is used in cell culture media formulations used in biomanufacturing. L-Histidine has been used to study cultures of the human T-lymphoblastic leukemia cell line MOLT-4 to study modulation of apoptosis.

Solubility
Freely soluble in water

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 5934-29-2
Molecular Formula C6H12ClN3O3
Molecular Weight (g/mol) 209.63
MDL Number MFCD00151027
InChI Key CMXXUDSWGMGYLZ-XRIGFGBMSA-N
Synonym l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
PubChem CID 165377
IUPAC Name (2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
SMILES O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O

Specifications

Melting Point ∽240°C (decomposition)
Color White
Beilstein 4168261
Merck Index 14,4720
Solubility Information Freely soluble in water
Quantity 10 g
Formula Weight 209.63 (191.62 Anhydrous)
Optical Rotation +10° (c=5 in 5N HCl)
Grade Cell Culture Reagent
Physical Form Powder
Chemical Name or Material L-Histidine monohydrochloride monohydrate
Show More Show Less
EINECSNumber 211-438-9
RTECSNumber MS3119000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only