missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine methyl ester dihydrochloride, 98+%

CAS: 7389-87-9 | C7H11ClN3O2 | 204.63 g/mol

Supplier:  Thermo Scientific Chemicals A1238614

Quantity 25 g

 View more versions of this product

Catalog No. AAA1238614

Only null left

Description

Description

L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Histidine methyl ester dihydrochloride is used to prepare optically pure L-(+)-ergothioneine.

Solubility
Soluble in dimethyl sulfoxide, methanol and water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is hygroscopic in nature. Store under dry inert gas.
Specifications

Chemical Identifiers

7389-87-9
204.63
VXXIPPPPKNWFLK-UHFFFAOYNA-N
2723645
C7H11ClN3O2
MFCD00012701
l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
[Cl].COC(=O)C(N)CC1=CN=CN1

Specifications

198°C (decomposition)
7389-87-9
MFCD00012701
l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
VXXIPPPPKNWFLK-UHFFFAOYNA-N
methyl (2S)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
204.63
242.11
≥98%
L-Histidine methyl ester dihydrochloride
C7H11ClN3O2
3572010
Soluble in dimethyl sulfoxide,methanol and water.
[Cl].COC(=O)C(N)CC1=CN=CN1
25 g
2723645
+10° (c=2 in water)
Hygroscopic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 230-973-9

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions