accessibility menu, dialog, popup

UserName

ClH3COONH2NHN

CAS RN 7389-87-9

IUPAC Name: methyl 2-amino-3-(1H-imidazol-5-yl)propanoate hydrochloridyl
Synonyms: methyl 2-amino-3-(3H-imidazol-4-yl)propanoate hydrochloridyl
Molecular Weight (g/mol): 204.63
Molecular Formula: C7H11ClN3O2
InChi Key: VXXIPPPPKNWFLK-UHFFFAOYNA-N
SMILES: [Cl].COC(=O)C(N)CC1=CN=CN1
Percent Purity 
  • (3)
  • (2)
  • (3)
Quantity 
  • (2)
  • (3)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (2)
  • (3)