missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Histidine methyl ester dihydrochloride, 97%

CAS: 7389-87-9 | C7H11ClN3O2 | 204.63 g/mol

$48.54 - $48.54

Chemical Identifiers

CAS 7389-87-9
Molecular Formula C7H11ClN3O2
Molecular Weight (g/mol) 204.63
MDL Number MFCD00012701
InChI Key VXXIPPPPKNWFLK-UHFFFAOYNA-N
Synonym l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
PubChem CID 2723645
IUPAC Name methyl (2S)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
SMILES [Cl].COC(=O)C(N)CC1=CN=CN1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC120820050
SDS View Documents
Thermo Scientific Chemicals
120820050
5 g Glass bottle
Each for $48.54
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7389-87-9
204.63
VXXIPPPPKNWFLK-UHFFFAOYNA-N
2723645
[Cl].COC(=O)C(N)CC1=CN=CN1
C7H11ClN3O2
MFCD00012701
l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
methyl (2S)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride

Specifications

7389-87-9
White
96% min. (Argentometry)
C7H11ClN3O2
5 g
+15° (24°C c=1,MeOH)
l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
[Cl].COC(=O)C(N)CC1=CN=CN1
204.63
242.11
Fine Crystalline Powder
207.0°C
Authentic
Glass bottle
MFCD00012701
25, 515
+ 15.00
VXXIPPPPKNWFLK-UHFFFAOYNA-N
methyl (2S)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
2723645
97%
L(+)-Histidine methyl ester dihydrochloride, 97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

EINECSNumber : 230-973-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only