missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Histidine methyl ester dihydrochloride, 97%

Catalog No. AC120820050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC120820050 5 g Glass bottle
AC120820250 25 g Glass bottle
AC120821000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC120820050 Supplier Thermo Scientific Chemicals Supplier No. 120820050
Only null left

CAS: 7389-87-9 | C7H11ClN3O2 | 204.63 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7389-87-9
Molecular Formula C7H11ClN3O2
Molecular Weight (g/mol) 204.63
MDL Number MFCD00012701
InChI Key VXXIPPPPKNWFLK-UHFFFAOYNA-N
Synonym l-histidine methyl ester dihydrochloride, h-his-ome.2hcl, methyl l-histidinate dihydrochloride, l-histidine, methyl ester, dihydrochloride, s-methyl 2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, l-+-histidine methyl ester dihydrochloride, methyl 2s-2-amino-3-1h-imidazol-4-yl propanoate dihydrochloride, methyl 2s-2-amino-3-3h-imidazol-4-yl propanoate dihydrochloride, histidine methyl ester dihydrochloride, c7h11n3o2.2hcl
PubChem CID 2723645
IUPAC Name methyl (2S)-2-amino-3-(1H-imidazol-5-yl)propanoate;dihydrochloride
SMILES [Cl].COC(=O)C(N)CC1=CN=CN1

Specifications

Melting Point 207.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Quantity 5 g
Beilstein 25, 515
Specific Rotation Condition +15° (24°C c=1,MeOH)
Specific Rotation + 15.00
Formula Weight 242.11
Percent Purity 97%
Physical Form Fine Crystalline Powder
Chemical Name or Material L(+)-Histidine methyl ester dihydrochloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
EINECSNumber 230-973-9

RUO – Research Use Only