missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine hydrochloride Solution, 100 mM Amino Acid in 0.1 M HCl, MilliporeSigma™ Supelco™

Catalog No. 111019663
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111019663 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111019663 Supplier Merck Emd Millipore Supplier No. 430115MLF

L-Histidine hydrochloride can undergo racemization due to heating in water, leading to the formation of DL-histidine. It can be used as a supplement in conditions of riboflavin vitamin deficiency.

Food and Beverages

Specifications

CAS 645-35-2
Linear Formula C6H9N3O2 · HCl
MDL Number MFCD00064556
Synonym L-Histidine monohydrochloride solution
Type Analytical Standard
Quantity 5 mL
Formula Weight 191.62
Grade Analytical Standard
Concentration or Composition (by Analyte or Components) 100 mM amino acid in 0.1 M HCl