missing translation for 'onlineSavingsMsg'
Learn More

L-Histidine hydrochloride monohydrate, 98%

CAS: 5934-29-2 | C6H12ClN3O3 | 209.63 g/mol

Supplier:  Thermo Scientific Chemicals 411730025

Quantity 2.5 kg
Packaging Plastic Bottle

 View more versions of this product

Catalog No. AC411730025


Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

5934-29-2
209.63
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
165377
O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
C6H12ClN3O3
MFCD00151027
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride

Specifications

L-Histidine hydrochloride monohydrate
200ppm max.
3 to 5 (2.5% soln. at 25°C)
10ppm max.
10ppm max.
98%
Plastic Bottle
MFCD00151027
25, 513
10, 4617
0.1% max.
O.Cl.N[C@@H](CC1=CNC=N1)C(O)=O
+ 9.90 (20.00°C c=11,6 N HCl)
209.63
209.64
Crystalline Powder or Crystals
98%
5934-29-2
White
Authentic
7% to 10% (3 hrs) (145 to 150°C)
0.5% max. (TLC)
C6H12ClN3O3
+ 9.90
300ppm max.
l-histidine hydrochloride hydrate, l-histidine hydrochloride monohydrate, s-2-amino-3-1h-imidazol-4-yl propanoic acid hydrochloride hydrate, h-his-oh.hcl.h2o, l-histidine monohydrochloride monohydrate, l-histidine, monohydrochloride, monohydrate, l-histidin hydrate hydrochloride, histidine, monohydrochloride, monohydrate, l, h-his-ohhclh2o, l-histidine, hydrochloride, monohydrate
CMXXUDSWGMGYLZ-XRIGFGBMSA-N
(2S)-2-amino-3-(1H-imidazol-4-yl)propanoic acid hydrate hydrochloride
2.5 kg
165377
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : MS3119000

SDS
Documents

Documents

Promotions

Promotions