missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Histidine, 98%

Catalog No. AC166150250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Plastic Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166150250 25 g Plastic Bottle
AC166151000 100 g Plastic Bottle
AC166155000 500 g Plastic Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC166150250 Supplier Thermo Scientific Chemicals Supplier No. 166150250

CAS: 71-00-1 | C6H9N3O2 | 155.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 71-00-1
Molecular Formula C6H9N3O2
Molecular Weight (g/mol) 155.16
MDL Number MFCD00064315
InChI Key HNDVDQJCIGZPNO-UHFFFAOYNA-N
Synonym l-histidine, histidine, h-his-oh, glyoxaline-5-alanine, anti-rheuma, l---histidine, istidina, s-histidine, l-histidin
PubChem CID 6274
ChEBI CHEBI:15971
IUPAC Name (2S)-2-amino-3-(1H-imidazol-5-yl)propanoic acid
SMILES NC(CC1=CN=CN1)C(O)=O

Specifications

Melting Point 282.0°C
Ammonium 200ppm max.
pH 7.0 to 8.5 (2% aq. soln.)
Color White or Yellow
Chloride 200ppm max.
Chlorine (Cl) 200ppm max.
Foreign Amino Acids 0.5% max. (TLC)
Heavy Metals (as Pb) 10ppm max.
Infrared Spectrum Authentic
Iron (Fe) 10ppm max.
Loss on Drying 0.2% max. (105°C, 3 hrs)
Assay Percent Range 98%
Packaging Plastic Bottle
Specific Rotation + 12.40
Beilstein 25, 513
Sulfate 200ppm max.
Merck Index 15, 4758
Transmittance 98% min. (c=5, H2O, 430nm) 1cm cell
Sulfated Ash 0.1% max.
Solubility Information Solubility in water: 41.6g/L (25 c). Other solubilities: soluble in mineral acids, insoluble in organic solvents
Specific Rotation Condition +12.40° (20°C c=11,6N HCl)
Quantity 25 g
Formula Weight 155.16
Percent Purity 98%
Physical Form Powder, Lumpy Powder or Melt
Chemical Name or Material L-Histidine
Show More Show Less

RUO – Research Use Only