missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Gulose 98.0+%, TCI America™
$125.13 - $1,840.46
Chemical Identifiers
| CAS | 6027-89-0 |
|---|---|
| Molecular Formula | C6H12O6 |
| Molecular Weight (g/mol) | 180.16 |
| MDL Number | MFCD00136022 |
| InChI Key | GZCGUPFRVQAUEE-JGWLITMVSA-N |
| Synonym | l-gulose, aldehydo-l-gulose, 2s,3s,4r,5s-2,3,4,5,6-pentahydroxyhexanal, unii-j96e9q45n7 |
| PubChem CID | 80127 |
| ChEBI | CHEBI:37701 |
| IUPAC Name | (2S,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal |
| SMILES | OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C=O |
Chemical Identifiers
| 6027-89-0 | |
| 180.16 | |
| GZCGUPFRVQAUEE-JGWLITMVSA-N | |
| 80127 | |
| (2S,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal |
| C6H12O6 | |
| MFCD00136022 | |
| l-gulose, aldehydo-l-gulose, 2s,3s,4r,5s-2,3,4,5,6-pentahydroxyhexanal, unii-j96e9q45n7 | |
| CHEBI:37701 | |
| OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C=O |
Specifications
| 6027-89-0 | |
| White | |
| MFCD00136022 | |
| l-gulose, aldehydo-l-gulose, 2s,3s,4r,5s-2,3,4,5,6-pentahydroxyhexanal, unii-j96e9q45n7 | |
| OC[C@H](O)[C@@H](O)[C@H](O)[C@H](O)C=O | |
| 180.16 | |
| CHEBI:37701 | |
| ≥98.0% (HPLC) | |
| L-Gulose |
| 132°C | |
| C6H12O6 | |
| 1 g | |
| GZCGUPFRVQAUEE-JGWLITMVSA-N | |
| (2S,3S,4R,5S)-2,3,4,5,6-pentahydroxyhexanal | |
| 80127 | |
| 180.16 | |
| Crystalline Powder |
Safety and Handling
TSCA : No