missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Gulonic Acid gamma-Lactone 98.0+%, TCI America™

Supplier:  TCI America G02355G

 View more versions of this product

Catalog No. G02355G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

L-(+)-Gulonic Acid gamma-Lactone
188°C
C6H10O6
5 g
SXZYCXMUPBBULW-SKNVOMKLSA-N
(3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
439373
178.14
Crystalline Powder
1128-23-0
White
MFCD00064331
l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone
C(C(C1C(C(C(=O)O1)O)O)O)O
178.14
CHEBI:17587
≥98.0% (GC)

Chemical Identifiers

1128-23-0
178.14
SXZYCXMUPBBULW-SKNVOMKLSA-N
439373
(3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
C6H10O6
MFCD00064331
l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone
CHEBI:17587
C(C(C1C(C(C(=O)O1)O)O)O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions