missing translation for 'onlineSavingsMsg'
Learn More

L-(+)-Gulonic Acid gamma-Lactone 98.0+%, TCI America™

Catalog No. p-7130202
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G02355G 5 g
G023525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G02355G Supplier TCI America Supplier No. G02355G
Only null left

Chemical Identifiers

CAS 1128-23-0
Molecular Formula C6H10O6
Molecular Weight (g/mol) 178.14
MDL Number MFCD00064331
InChI Key SXZYCXMUPBBULW-SKNVOMKLSA-N
Synonym l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone
PubChem CID 439373
ChEBI CHEBI:17587
IUPAC Name (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one
SMILES C(C(C1C(C(C(=O)O1)O)O)O)O

Specifications

Melting Point 188°C
Color White
Quantity 5 g
Formula Weight 178.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material L-(+)-Gulonic Acid gamma-Lactone