missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Gulonic acid-1,4-lactone, 95%
CAS: 1128-23-0 | C6H10O6 | 178.14 g/mol
Supplier: Thermo Scientific Chemicals J6693414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| L-Gulonic acid-1,4-lactone | |
| 184°C to 190°C | |
| C6H10O6 | |
| 25 g | |
| l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone | |
| SXZYCXMUPBBULW-SKNVOMKLSA-N | |
| C(C(C1C(C(C(=O)O1)O)O)O)O | |
| 178.14 | |
| CHEBI:17587 | |
| 95% |
| 1128-23-0 | |
| White | |
| MFCD00064331 | |
| 83002 | |
| Soluble in water | |
| +54.5° (c=4 in Water) | |
| (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one | |
| 439373 | |
| 178.14 | |
| Powder |
Chemical Identifiers
| 1128-23-0 | |
| 178.14 | |
| SXZYCXMUPBBULW-SKNVOMKLSA-N | |
| 439373 | |
| (3S,4R,5R)-5-[(1S)-1,2-dihydroxyethyl]-3,4-dihydroxyoxolan-2-one |
| C6H10O6 | |
| MFCD00064331 | |
| l-gulonolactone, l-gulono-1,4-lactone, l-gulonic acid gamma-lactone, 3s,4r,5r-5-s-1,2-dihydroxyethyl-3,4-dihydroxydihydrofuran-2 3h-one, l-gulono-gamma-lactone, gamma-gulonolactone, l-gulonic gamma-lactone, l-+-gulono-1,4-lactone, l +-gulonic acid gamma-lactone, l-+-gulonic acid gamma-lactone | |
| CHEBI:17587 | |
| C(C(C1C(C(C(=O)O1)O)O)O)O |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only