Learn More
L-Glutamine, 99%
Involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. | CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
Supplier: Thermo Scientific Chemicals A1420130
Description
L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsL-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.
Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in methanol, ether, benzene, acetone, ethyl acetate and chloroform.
Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture and strong oxidizing agents.
Specifications
| 185°C (decomposition) | |
| 56-85-9 | |
| Plastic Bottle | |
| 1723797 | |
| Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in methanol,ether,benzene,acetone,ethyl acetate and chloroform. | |
| N[C@@H](CCC(N)=O)C(O)=O | |
| 146.15 | |
| 5961 | |
| 146.15 | |
| Powder |
| L-Glutamine | |
| C5H10N2O3 | |
| MFCD00008044 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| (2S)-2-amino-4-carbamoylbutanoic acid | |
| 250 g | |
| CHEBI:18050 | |
| 99% |
Chemical Identifiers
| 56-85-9 | |
| 146.15 | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| 5961 | |
| N[C@@H](CCC(N)=O)C(O)=O |
| C5H10N2O3 | |
| MFCD00008044 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| CHEBI:18050 |
Safety and Handling
P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H320-H335-H336
EINECSNumber : 200-292-1
RTECSNumber : MA2275100
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only