missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamine, 99%

Catalog No. p-7076512
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1420118 50 g
AAA1420130 250 g
AAA142010B 1000 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1420118 Supplier Thermo Scientific Chemicals Supplier No. A1420118
Only null left

Involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. | CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamine is involved in many metabolic processes and regulates acid-base balance in the kidney. It is an important amino acid for the incorporation of ammonium ions into biomolecules. It is a vital component of culture media that serves as a major energy source for cells in culture. As a component in oral supplementation, it is useful for patients who have advanced stages of cancer to reverse cachexia (i.e. muscle wasting). It is also used for attention deficit-hyperactivity disorder (ADHD), a urinary condition called cystinuria, sickle cell anemia and for alcohol withdrawal support. It finds applications in patients with HIV (AIDS) to prevent weight loss.

Solubility
Soluble in water, dimethyl sulfoxide and ethanol. Insoluble in methanol, ether, benzene, acetone, ethyl acetate and chloroform.

Notes
Moisture and light sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with moisture and strong oxidizing agents.

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
SMILES N[C@@H](CCC(N)=O)C(O)=O

Specifications

Melting Point 185°C (decomposition)
Assay Percent Range 99%
Beilstein 1723797
Solubility Information Soluble in water,dimethyl sulfoxide and ethanol. Insoluble in methanol,ether,benzene,acetone,ethyl acetate and chloroform.
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
Quantity 50 g
Formula Weight 146.15
Chemical Name or Material L-Glutamine
Hazard Category H315-H319-H320-H335-H336
Precautionary Statement P261-P264b-P271-P280g-P280i-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 200-292-1
RTECSNumber MA2275100
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only