missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamine, 99%

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

Supplier:  Thermo Scientific Chemicals 119951000

 View more versions of this product

Catalog No. AC119951000

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

185.0°C
99%
4 to 6 (2.5% soln. at 25°C)
200ppm max.
Authentic
0.2% max. (105°C, 3 hrs)
0.5% max. (TLC)
Plastic Bottle
MFCD00008044
04, 491
95% min. (c=10, 2N HCl, 430nm) 1cm cell
0.1% max.
Solubility in water: 35g/L (20°C). Other solubilities: practically insoluble in methanol, ethanol, ether,, benzene, acetone, ethyl acetate, chloroform, soluble in mineral acids, soluble in ethanol: 46 μg/100mL (25°C), soluble in methanol: 3.5 mg/100mL (2
N[C@@H](CCC(N)=O)C(O)=O
146.15
5961
146.15
Crystalline Powder or Crystals
L(+)-Glutamine
56-85-9
White
10ppm max.
10ppm max.
99%
C5H10N2O3
H2NCOCH2CH2CH(NH2)CO2H
+31.5° to +33° (20°C, 589nm) (c=10, 2N HCl)
200ppm max.
15, 4507
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
ZDXPYRJPNDTMRX-VKHMYHEASA-N
(2S)-2,5-diamino-5-oxopentanoic acid
100 g
CHEBI:18050
99%

Chemical Identifiers

56-85-9
146.15
ZDXPYRJPNDTMRX-VKHMYHEASA-N
5961
(2S)-2,5-diamino-5-oxopentanoic acid
C5H10N2O3
MFCD00008044
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
CHEBI:18050
N[C@@H](CCC(N)=O)C(O)=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

LIGHT SENSITIVE

EINECSNumber : 200-292-1

RTECSNumber : MA2275100

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only