missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L(+)-Glutamine, 99%

CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol

$143.44 - $2,157.00

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2,5-diamino-5-oxopentanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC119951000
View Documents Promotion Details
Thermo Scientific Chemicals
119951000
100 g Plastic Bottle
Each for $143.44
Only null left
 
AC119955000
View Documents Promotion Details
Thermo Scientific Chemicals
119955000
500 g Plastic Bottle
Each for $514.97
Only null left
 
AC119950025
View Documents Promotion Details
Thermo Scientific Chemicals
119950025
2.5 kg Plastic Bottle
Each for $2,157.00
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56-85-9
146.15
ZDXPYRJPNDTMRX-VKHMYHEASA-N
5961
(2S)-2,5-diamino-5-oxopentanoic acid
C5H10N2O3
MFCD00008044
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
CHEBI:18050
N[C@@H](CCC(N)=O)C(O)=O

Specifications

185.0°C
4 to 6 (2.5% soln. at 25°C)
200ppm max.
Authentic
0.2% max. (105°C, 3 hrs)
0.5% max. (TLC)
H2NCOCH2CH2CH(NH2)CO2H
04, 491
200ppm max.
95% min. (c=10, 2N HCl, 430nm) 1cm cell
0.1% max.
ZDXPYRJPNDTMRX-VKHMYHEASA-N
(2S)-2,5-diamino-5-oxopentanoic acid
5961
146.15
Crystalline Powder or Crystals
56-85-9
White
10ppm max.
10ppm max.
99%
C5H10N2O3
MFCD00008044
+31.5° to +33° (20°C, 589nm) (c=10, 2N HCl)
15, 4507
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
Solubility in water: 35g/L (20°C). Other solubilities: practically insoluble in methanol, ethanol, ether,, benzene, acetone, ethyl acetate, chloroform, soluble in mineral acids, soluble in ethanol: 46 μg/100mL (25°C), soluble in methanol: 3.5 mg/100mL (2
N[C@@H](CCC(N)=O)C(O)=O
146.15
CHEBI:18050
99%
L(+)-Glutamine, 99%
Videos
Safety and Handling

Safety and Handling

MOISTURE SENSITIVE

LIGHT SENSITIVE

EINECSNumber : 200-292-1

RTECSNumber : MA2275100

TSCA : TSCA

SDS
Documents

Documents

RUO – Research Use Only