Learn More
Thermo Scientific Chemicals L(+)-Glutamine, 99%
CAS: 56-85-9 | C5H10N2O3 | 146.15 g/mol
Supplier: Thermo Scientific Chemicals 119950025
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| 185.0°C | |
| 99% | |
| 4 to 6 (2.5% soln. at 25°C) | |
| 200ppm max. | |
| Authentic | |
| 0.2% max. (105°C, 3 hrs) | |
| 0.5% max. (TLC) | |
| Plastic Bottle | |
| MFCD00008044 | |
| 04, 491 | |
| 95% min. (c=10, 2N HCl, 430nm) 1cm cell | |
| 0.1% max. | |
| Solubility in water: 35g/L (20°C). Other solubilities: practically insoluble in methanol, ethanol, ether,, benzene, acetone, ethyl acetate, chloroform, soluble in mineral acids, soluble in ethanol: 46 μg/100mL (25°C), soluble in methanol: 3.5 mg/100mL (2 | |
| N[C@@H](CCC(N)=O)C(O)=O | |
| 146.15 | |
| 5961 | |
| 146.15 | |
| Crystalline Powder or Crystals |
| L(+)-Glutamine | |
| 56-85-9 | |
| White | |
| 10ppm max. | |
| 10ppm max. | |
| 99% | |
| C5H10N2O3 | |
| H2NCOCH2CH2CH(NH2)CO2H | |
| +31.5° to +33° (20°C, 589nm) (c=10, 2N HCl) | |
| 200ppm max. | |
| 15, 4507 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| (2S)-2,5-diamino-5-oxopentanoic acid | |
| 2.5 kg | |
| CHEBI:18050 | |
| 99% |
Chemical Identifiers
| 56-85-9 | |
| 146.15 | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| 5961 | |
| (2S)-2,5-diamino-5-oxopentanoic acid |
| C5H10N2O3 | |
| MFCD00008044 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| CHEBI:18050 | |
| N[C@@H](CCC(N)=O)C(O)=O |
Safety and Handling
MOISTURE SENSITIVE
LIGHT SENSITIVE
EINECSNumber : 200-292-1
RTECSNumber : MA2275100
TSCA : TSCA
RUO – Research Use Only