missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
L-Glutamine, 99-100%, MP Biomedicals
Manufacturer: Mp Biomedicals 0219467880
Specifications
L-Glutamine, 99-100% | |
C5H10N2O3 | |
ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
(2S)-2,5-diamino-5-oxopentanoic acid | |
5961 | |
Crystalline | |
100g |
56-85-9 | |
cebrogen, glumin, glutamic acid amide, glutamine, h-gln-oh, l-+-glutamine, l-glutamine, levoglutamid, levoglutamide, stimulina | |
C(CC(=O)N)C(C(=O)O)N | |
146.146 | |
CHEBI:18050 | |
Cell Culture Reagent |
Chemical Identifiers
56-85-9 | |
146.146 | |
cebrogen, glumin, glutamic acid amide, glutamine, h-gln-oh, l-+-glutamine, l-glutamine, levoglutamid, levoglutamide, stimulina | |
CHEBI:18050 | |
C(CC(=O)N)C(C(=O)O)N |
C5H10N2O3 | |
ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
5961 | |
(2S)-2,5-diamino-5-oxopentanoic acid |
Safety and Handling
Recommended Storage : Room temperature