missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamine, 99-100%, MP Biomedicals
$144.56
Chemical Identifiers
| CAS | 56-85-9 |
|---|---|
| Molecular Formula | C5H10N2O3 |
| Molecular Weight (g/mol) | 146.15 |
| MDL Number | MFCD00008044 |
| InChI Key | ZDXPYRJPNDTMRX-VKHMYHEASA-N |
| Synonym | l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid |
| PubChem CID | 5961 |
| ChEBI | CHEBI:18050 |
| IUPAC Name | (2S)-2-amino-4-carbamoylbutanoic acid |
| SMILES | N[C@@H](CCC(N)=O)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ICN19467880
|
N/A | 100 g |
Each for $144.56
|
|
|||||
Chemical Identifiers
| C5H10N2O3 | |
| MFCD00008044 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| CHEBI:18050 | |
| N[C@@H](CCC(N)=O)C(O)=O |
Specifications
| 56-85-9 | |
| MFCD00008044 | |
| ZDXPYRJPNDTMRX-VKHMYHEASA-N | |
| (2S)-2-amino-4-carbamoylbutanoic acid | |
| 5961 | |
| Cell Culture | |
| L-Glutamine, 99-100% |
| C5H10N2O3 | |
| l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid | |
| N[C@@H](CCC(N)=O)C(O)=O | |
| 146.15 | |
| CHEBI:18050 | |
| Crystals |
Safety and Handling
Recommended Storage : Room temperature