missing translation for 'onlineSavingsMsg'
Learn More

L-glutamine, 99 to 100%, MP Biomedicals™

$123.86 - $290.44

Chemical Identifiers

CAS 56-85-9
Molecular Formula C5H10N2O3
Molecular Weight (g/mol) 146.15
MDL Number MFCD00008044
InChI Key ZDXPYRJPNDTMRX-VKHMYHEASA-N
Synonym l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
PubChem CID 5961
ChEBI CHEBI:18050
IUPAC Name (2S)-2-amino-4-carbamoylbutanoic acid
SMILES N[C@@H](CCC(N)=O)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
ICN10180680
View Documents Promotion Details
N/A 100 g
Each for $123.86
Only null left
 
ICN10180683
View Documents Promotion Details
N/A 250 g
Each for $290.44
Only null left
 
Description

Description

Specifications

Chemical Identifiers

56-85-9
146.15
ZDXPYRJPNDTMRX-VKHMYHEASA-N
5961
(2S)-2-amino-4-carbamoylbutanoic acid
C5H10N2O3
MFCD00008044
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
CHEBI:18050
N[C@@H](CCC(N)=O)C(O)=O

Specifications

56-85-9
99 to 100%
MFCD00008044
ZDXPYRJPNDTMRX-VKHMYHEASA-N
(2S)-2-amino-4-carbamoylbutanoic acid
5961
146.1
L-Glutamine
White
C5H10N2O3
l-glutamine, glutamine, levoglutamide, l-+-glutamine, glutamic acid amide, h-gln-oh, stimulina, cebrogen, glumin, levoglutamid
N[C@@H](CCC(N)=O)C(O)=O
146.15
CHEBI:18050
Powder
Videos
SDS
Documents

Documents