missing translation for 'onlineSavingsMsg'
Learn More

Cytiva L-Glutamine

Catalog No. SH3003401
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
SH3003401 100 mL
SH3003402 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. SH3003401 Supplier Cytiva Supplier No. SH30034.01
NLSU: Get 1 Consumable Product When You Buy 1 Obtenez 1 produit consommable à l’achat d’un produit
View all promotions
Edge

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Supplement serum- and protein-free media using this amino acid.

  • Format: Liquid

Specifications

Content And Storage Store frozen
Description L-Glutamine, 200mM Solution (29.2mg/mL-Glutamine in 0.85% NaCl), 100mL
Concentration 29.2 mg/mL
Form Liquid
Sterilization Method 0.1 μm Sterile-filtered
Quantity 100 mL