missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid Monopotassium Salt, Spectrum™ Chemical
G1090, 6382-01-0, C5H8KNO4·H2O
$146.90 - $1,562.50
Chemical Identifiers
| CAS | 6382-01-0 |
|---|---|
| Molecular Formula | C5H10KNO5 |
| Molecular Weight (g/mol) | 203.24 |
| InChI Key | XIBUKSQTWSKJMQ-QTNFYWBSSA-M |
| IUPAC Name | potassium (4S)-4-amino-4-carboxybutanoate hydrate |
| SMILES | O.[K+].N[C@@H](CCC([O-])=O)C(O)=O |
Description
Spectrum™ Chemical L-Glutamic Acid Monopotassium Salt , also known as sodium glutamate, is the sodium salt of glutamic acid. It is used in the food industry most usually as a flavor with an umami taste. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.Chemical Identifiers
| 6382-01-0 | |
| 203.24 | |
| potassium (4S)-4-amino-4-carboxybutanoate hydrate |
| C5H10KNO5 | |
| XIBUKSQTWSKJMQ-QTNFYWBSSA-M | |
| O.[K+].N[C@@H](CCC([O-])=O)C(O)=O |
Specifications
| 6382-01-0 | |
| C5H10KNO5 | |
| O.[K+].N[C@@H](CCC([O-])=O)C(O)=O | |
| 203.24 | |
| L-Glutamic Acid Monopotassium Salt |
| 100% | |
| XIBUKSQTWSKJMQ-QTNFYWBSSA-M | |
| potassium (4S)-4-amino-4-carboxybutanoate hydrate | |
| Technical |