missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid Hydrochloride 98.0+%, TCI America™
$32.28 - $98.72
Chemical Identifiers
| CAS | 138-15-8 |
|---|---|
| Molecular Formula | C5H10ClNO4 |
| Molecular Weight (g/mol) | 183.588 |
| MDL Number | MFCD00012619 |
| InChI Key | RPAJSBKBKSSMLJ-DFWYDOINSA-N |
| Synonym | l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin |
| PubChem CID | 2723891 |
| IUPAC Name | (2S)-2-aminopentanedioic acid;hydrochloride |
| SMILES | C(CC(=O)O)C(C(=O)O)N.Cl |
Chemical Identifiers
| C5H10ClNO4 | |
| MFCD00012619 | |
| l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
| (2S)-2-aminopentanedioic acid;hydrochloride |
Specifications
| 138-15-8 | |
| C5H10ClNO4 | |
| l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin | |
| C(CC(=O)O)C(C(=O)O)N.Cl | |
| 183.588 | |
| 183.59 | |
| Crystalline Powder |
| White | |
| MFCD00012619 | |
| RPAJSBKBKSSMLJ-DFWYDOINSA-N | |
| (2S)-2-aminopentanedioic acid;hydrochloride | |
| 2723891 | |
| ≥98.0% (T) | |
| L-Glutamic Acid Hydrochloride |
Safety and Handling
TSCA : Yes