missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid Hydrochloride 98.0+%, TCI America™

Catalog No. G006025G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G006025G 25 g
G0060500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G006025G Supplier TCI America Supplier No. G006025G

Chemical Identifiers

CAS 138-15-8
Molecular Formula C5H10ClNO4
Molecular Weight (g/mol) 183.588
MDL Number MFCD00012619
InChI Key RPAJSBKBKSSMLJ-DFWYDOINSA-N
Synonym l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin
PubChem CID 2723891
IUPAC Name (2S)-2-aminopentanedioic acid;hydrochloride
SMILES C(CC(=O)O)C(C(=O)O)N.Cl

Specifications

Color White
Quantity 25 g
Formula Weight 183.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material L-Glutamic Acid Hydrochloride