missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals L-Glutamic acid hydrochloride, 99%

CAS: 138-15-8 | C5H10ClNO4 | 183.588 g/mol

Supplier:  Thermo Scientific Chemicals A125050B

Quantity 1000 g

 View more versions of this product

Catalog No. AAA125050B


Only null left

Description

Description

L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid hydrochloride is used as a digestive aid. It is used in cell culture as a component of MEM non-essential amino acids solution. It has been used as a nitrogen source in the culture of Aspergillus fumigatus NRRL 2436 for fumagillin production.

Solubility
Soluble in water (490 g/L).

Notes
Hygroscopic. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

138-15-8
183.588
RPAJSBKBKSSMLJ-DFWYDOINSA-N
2723891
C(CC(=O)O)C(C(=O)O)N.Cl
C5H10ClNO4
MFCD00012619
l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin
(2S)-2-aminopentanedioic acid;hydrochloride

Specifications

203°C (decomposition)
138-15-8
MFCD00012619
14,4469
Soluble in water (490g/L).
1.525
(2S)-2-aminopentanedioic acid;hydrochloride
183.588
183.59
99%
L-Glutamic acid hydrochloride
C5H10ClNO4
3565569
l-glutamic acid hydrochloride, s-2-aminopentanedioic acid hydrochloride, l-+-glutamic acid hydrochloride, glutamic acid hydrochloride, unii-m0c2sp444t, glu hcl, acidogen, acidulin, gastuloric, glutamidin
RPAJSBKBKSSMLJ-DFWYDOINSA-N
C(CC(=O)O)C(C(=O)O)N.Cl
1000 g
2723891
+23° (c=6 in Water)
Hygroscopic
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H315
Causes serious eye damage.
May cause respiratory irritation.
Causes skin irritation.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

EINECSNumber : 205-315-9

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions