missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic Acid, FCC, 98.5-101.5%, Spectrum™ Chemical
G1036, 56-86-0, C5H9NO4
Supplier: Spectrum Chemical Mfg Cor G103625KGBL
Description
Spectrum™ Chemical L-Glutamic Acid, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 56-86-0 | |
| 0.3% | |
| Fiber Drum | |
| MFCD00002634 | |
| WHUUTDBJXJRKMK-UHFFFAOYNA-N | |
| NC(CCC(O)=O)C(O)=O | |
| 147.13 | |
| FCC |
| 100% | |
| 0.1% | |
| C5H9NO4 | |
| 25 kg | |
| +31.5° to +32.5° | |
| 2-aminopentanedioic acid | |
| 98.5 to 101.5% |
Chemical Identifiers
| 56-86-0 | |
| 147.13 | |
| WHUUTDBJXJRKMK-UHFFFAOYNA-N | |
| NC(CCC(O)=O)C(O)=O |
| C5H9NO4 | |
| MFCD00002634 | |
| 2-aminopentanedioic acid |