missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, FCC, 98.5-101.5%, Spectrum™ Chemical

Catalog No. p-7157484
Change view
Click to view available options
Quantity:
100 g
1 kg
12 kg
25 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18603830 100 g Poly Bottle
18603831 1 kg Poly Bottle
18603834 12 kg Poly Pail
18603832 25 kg Fiber Drum
18603833 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18603830 Supplier Spectrum Chemical Mfg Cor Supplier No. G1036100GM

G1036, 56-86-0, C5H9NO4

Spectrum™ Chemical L-Glutamic Acid, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 56-86-0
Molecular Formula C5H9NO4
Molecular Weight (g/mol) 147.13
MDL Number MFCD00002634
InChI Key WHUUTDBJXJRKMK-UHFFFAOYNA-N
IUPAC Name 2-aminopentanedioic acid
SMILES NC(CCC(O)=O)C(O)=O

Specifications

CAS Max % 100%
Ignition Residue 0.3%
Loss on Drying 0.1%
Packaging Poly Bottle
Quantity 100 g
Optical Rotation +31.5° to +32.5°
Percent Purity 98.5 to 101.5%
Grade FCC