missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid dimethyl ester hydrochloride, 99%, pure, Thermo Scientific™

$115.70 - $490.85

Chemical Identifiers

CAS 23150-65-4
Molecular Formula C7H13ClNO4
Molecular Weight (g/mol) 210.63
MDL Number MFCD00038879
InChI Key BPHCSYSXOKTCMA-UHFFFAOYNA-N
Synonym l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride
PubChem CID 12917568
IUPAC Name dimethyl (2S)-2-aminopentanedioate;hydrochloride
SMILES [Cl].COC(=O)CCC(N)C(=O)OC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC441940100
View Documents Promotion Details
N/A 10g
Each for $115.70
Only null left
 
AC441940500
View Documents Promotion Details
N/A 50g
Each for $490.85
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

23150-65-4
210.63
BPHCSYSXOKTCMA-UHFFFAOYNA-N
12917568
[Cl].COC(=O)CCC(N)C(=O)OC
C7H13ClNO4
MFCD00038879
l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride
dimethyl (2S)-2-aminopentanedioate;hydrochloride

Specifications

23150-65-4
98.5% min. (Argentometry)
C7H13ClNO4
26
BPHCSYSXOKTCMA-UHFFFAOYNA-N
dimethyl (2S)-2-aminopentanedioate;hydrochloride
12917568
99%
L-Glutamic acid dimethyl ester hydrochloride
Authentic
Glass bottle
MFCD00038879
l-glutamic acid dimethyl ester hydrochloride, h-glu ome-ome.hcl, dimethyl l-glutamate hydrochloride, s-dimethyl 2-aminopentanedioate hydrochloride, unii-185vc5x3pf, 1,5-dimethyl 2s-2-aminopentanedioate hydrochloride, h-glu ome-ome inverted exclamation mark currencyhcl, s-dimethyl 2-aminopentanedioate hcl, glutamic acid dimethyl ester hydrochloride, dimethyl glutamic acid hydrochloride
[Cl].COC(=O)CCC(N)C(=O)OC
210.63
211.65
Pure
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 245-461-

SDS
Documents

Documents

RUO – Research Use Only