missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid dibenzyl ester hydrochloride, 98%

CAS: 4561-10-8 | C19H22ClNO4 | 363.84 g/mol

$87.22 - $291.27

Chemical Identifiers

CAS 4561-10-8
MDL Number MFCD00035103
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6359803
View Documents
Thermo Scientific Chemicals
H6359803
1 g
Each for $87.22
Only null left
 
AAH6359806
View Documents
Thermo Scientific Chemicals
H6359806
5 g
Each for $291.27
Only null left
 
Description

Description

L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
L-Glutamic acid dibenzyl ester hydrochloride oxalate used as a intermediate in pharmaceutical and chemical synthesis. Also used in medicine research.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

4561-10-8
MFCD00035103

Specifications

4561-10-8
C19H22ClNO4
s-dibenzyl 2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hcl, l-glutamic acid dibenzyl ester hydrochloride, l-glutamic acid, bis phenylmethyl ester, hydrochloride, s-dibenzyl 2-aminopentanedioate hcl, dibenzyl l-glutaminate hydrochloride, glutamic acid, dibenzyl ester, hydrochloride, l, 1,5-dibenzyl 2s-2-aminopentanedioate hydrochloride, h-glu obzl-obzl.hci, h-glu obzl-obzl hcl
C1=CC=C(C=C1)COC(=O)CCC(C(=O)OCC2=CC=CC=C2)N.Cl
1 g
199613
98%
L-Glutamic acid dibenzyl ester hydrochloride
White
MFCD00035103
GRGJVECUQLAEDM-LMOVPXPDSA-N
dibenzyl (2S)-2-aminopentanedioate;hydrochloride
363.84
363.84
Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : MA0885000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only