accessibility menu, dialog, popup

UserName

CAS RN 4561-10-8

ClNH2(S)OOOOH

CAS RN 4561-10-8

IUPAC Name: hydrogen 1,5-dibenzyl (2S)-2-aminopentanedioate chloride
Synonyms: hydrogen 1,5-dibenzyl (2S)-2-aminopentanedioate chloride
Molecular Weight (g/mol): 363.84
Molecular Formula: C19H22ClNO4
InChi Key: GRGJVECUQLAEDM-LMOVPXPDSA-N
SMILES: [H+].[Cl-].N[C@@H](CCC(=O)OCC1=CC=CC=C1)C(=O)OCC1=CC=CC=C1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)