missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic Acid, 99+%, Monosodium Salt, MP Biomedicals

Catalog No. ICN19467780
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19467780 100 g
ICN19467790 500 g
ICN19467791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN19467780 Supplier MP Biomedicals Inc Supplier No. 0219467780

Chemical Identifiers

CAS 142-47-2
Molecular Formula C5H8NNaO4
Molecular Weight (g/mol) 169.11
MDL Number MFCD00150138
InChI Key LPUQAYUQRXPFSQ-UHFFFAOYNA-M
Synonym natriumglutaminat, sodium glutamate, glutamate sodium, monosodioglutammato, sodium l-glutamate, glutammato monosodico, monosodium l-glutamate, sodium hydrogen glutamate, monosodium glutamate, glutamic acid, monosodium salt
PubChem CID 86748263
IUPAC Name sodium 2-amino-4-carboxybutanoate
SMILES [Na+].NC(CCC(O)=O)C([O-])=O

Specifications

Quantity 100 g
Formula Weight 169.1
Grade Cell Culture
Chemical Name or Material L-Glutamic Acid, 99+%, Monosodium Salt
Recommended Storage Room temperature