missing translation for 'onlineSavingsMsg'
Learn More
Learn More
L-Glutamic acid, 98.5 to 101.5% (Dry Basis), Ultrapure
CAS: 56-86-0 | C5H9NO4 | 147.13 g/mol
$169.58 - $1,534.92
Chemical Identifiers
| CAS | 56-86-0 |
|---|---|
| Molecular Formula | C5H9NO4 |
| Molecular Weight (g/mol) | 147.13 |
| MDL Number | MFCD00002634 |
| InChI Key | WHUUTDBJXJRKMK-VKHMYHEASA-N |
| Synonym | l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex |
| PubChem CID | 33032 |
| ChEBI | CHEBI:16015 |
| IUPAC Name | (2 S)-2-aminopentanedioic acid |
| SMILES | C(CC(=O)O)C(C(=O)O)N |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AAJ16215A1
|
N/A | 1 kg | Plastic Bottle |
Each for $169.58
|
|
||||
|
AAJ16215C4
|
N/A | 25 kg | Plastic Bottle |
Each for $1,534.92
|
|
||||
Chemical Identifiers
| C5H9NO4 | |
| MFCD00002634 | |
| l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex | |
| CHEBI:16015 | |
| C(CC(=O)O)C(C(=O)O)N |
Specifications
| 189°C to 192°C (decomposition) | |
| C5H9NO4 | |
| MFCD00002634 | |
| l-glutamic acid, glutamic acid, 2 s-2-aminopentanedioic acid, h-glu-oh, s-2-aminopentanedioic acid, glutaminol, l-glutaminic acid, glutacid, glutamicol, glutamidex | |
| C(CC(=O)O)C(C(=O)O)N | |
| (2 S)-2-aminopentanedioic acid | |
| 33032 | |
| 147.13 | |
| 1.525 | |
| Dry basis |
| 56-86-0 | |
| HOOCCH2CH2CH(NH2)COOH | |
| 1723801 | |
| WHUUTDBJXJRKMK-VKHMYHEASA-N | |
| 1.525 | |
| 147.13 | |
| CHEBI:16015 | |
| 98.5 to 101.5% | |
| Crystalline Powder | |
| L-Glutamic acid |
Safety and Handling
H500
EINECSNumber : 200-293-7
RTECSNumber : LZ9700000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only