missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-benzyl ester, 99%

CAS: 1676-73-9 | C12H15NO4 | 237.255 g/mol

$53.97 - $499.66

Chemical Identifiers

CAS 1676-73-9
Molecular Formula C12H15NO4
Molecular Weight (g/mol) 237.255
MDL Number MFCD00002633
InChI Key BGGHCRNCRWQABU-JTQLQIEISA-N
Synonym h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
PubChem CID 122337
IUPAC Name (2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2218403
View Documents
Thermo Scientific Chemicals
B2218403
1 g
Each for $53.97
Only null left
 
AAB2218406
View Documents
Thermo Scientific Chemicals
B2218406
5 g
Each for $142.41
Only null left
 
AAB2218414
View Documents
Thermo Scientific Chemicals
B2218414
25 g
Each for $499.66
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1676-73-9
237.255
BGGHCRNCRWQABU-JTQLQIEISA-N
122337
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
C12H15NO4
MFCD00002633
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid

Specifications

162°C to 165°C
C12H15NO4
1885646
BGGHCRNCRWQABU-JTQLQIEISA-N
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
237.255
+23° (c=1 in Acetic Acid)
99%
1676-73-9
MFCD00002633
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
1 g
122337
237.26
L-Glutamic acid 5-benzyl ester
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-826-1

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only