missing translation for 'onlineSavingsMsg'
Learn More

L-Glutamic acid 5-benzyl ester, 99%

CAS: 1676-73-9 | C12H15NO4 | 237.255 g/mol

Supplier:  Thermo Scientific Chemicals B2218406

 View more versions of this product

Catalog No. AAB2218406


Only null left

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

162°C to 165°C
1676-73-9
MFCD00002633
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
5 g
122337
237.26
L-Glutamic acid 5-benzyl ester
C12H15NO4
1885646
BGGHCRNCRWQABU-JTQLQIEISA-N
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
237.255
+23° (c=1 in Acetic Acid)
99%

Chemical Identifiers

1676-73-9
237.255
BGGHCRNCRWQABU-JTQLQIEISA-N
122337
C1=CC=C(C=C1)COC(=O)CCC(C(=O)O)N
C12H15NO4
MFCD00002633
h-glu obzl-oh, gamma-benzyl l-glutamate, l-glutamic acid 5-benzyl ester, 5-benzyl l-glutamate, l-glutamic acid gamma-benzyl ester, s-2-amino-5-benzyloxy-5-oxopentanoic acid, 2s-2-amino-5-benzyloxy-5-oxopentanoic acid, glutamic acid, 5-benzyl ester, l, gama-benzyl l-glutamate, h-glu obzl-oh.hcl
(2S)-2-amino-5-oxo-5-phenylmethoxypentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 216-826-1

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only